Zonal Rotor without core, Titanium, Ti-15, Optima
Catalog # 521-4069
Supplier: Beckman Coulter
Specifications
- Désignation:Ti-15 zonal rotor without core, titanium. Major applications: Rate-zonal centrifugation of subcellular particles
- Cdt:1
- Pour:Speciality (ultracentrifugation)
- Max. RPM:32000