Fixed-angle aluminium rotor, Avanti JA-25.15
Référence 521-3995
Fournisseur: Beckman Coulter
Spécifications
- Capacité:360 ml
- Désignation:JA-25.15 fixed angle (high performance) aluminium rotor with single locked lid, 24×15 ml, 25000 rpm, 74200×g, for Avanti J Series centrifuges (except Avanti J-E) and J2 Series centrifuges
- Cdt:1
- Pour:Major applications: Harvesting bacteria, processing tissue homogenates, subcellular particulates, routine pelleting such as precipitates and phase separations
- Max. RPM:25000
- No. of tubes accepted:24×15 ml/18×100 mm (24×15 ml/1×4")
- Radius:outer row 106 inner row 86