Swinging-bucket titanium rotor package/assembly, TLS-55
Fournisseur: Beckman Coulter
Certificats
À propos de cet article
Major applications: RNA pelleting in 2 to 3 hours; subcellular fractionation in sucrose gradients; protein separations in sucrose gradients.
Note: Non-precipitating solutions up to 1,7 g/ml in density can be run on this rotor without a reduction in rotor speed.
Spécifications
- Catalog No:
- 521-3802
- 521-3804
- Capacité:
- 8,8 ml
- 8,8 ml
- No. of tubes accepted:
- 4×2,2ml / 11×34 mm
- 4×2,2ml / 11×34 mm
- Radius:
- 76,5 mm
- 76,5 mm
- Max. RPM:
- 55000
- 55000