UA tape
N° de catalogue 300-0565
Fournisseur: Sampling Systems
Spécifications
- Largeur:50 mm
- Désignation:UA tape
- Longueur:50 m
- Cdt:1
Spécifications
À propos de cet article
UA tape is a highly versatile solution for many applications.
- Easy to write on
- Ideal for use in production, warehouses, and laboratories
UA Tape offers the ultimate in adhesive strength. It is ideal for sealing sacks and bags after sampling and dispensing. The tape is also foil backed to provide extra strength to the label.