Analytical 8-place titanium rotor, An-50 Ti
521-4025
- Désignation:An-50 Ti analytical 8-place titanium rotor, requires XL-A/XL-I user interface
- Cdt:1
- Pour:Optima AUC and Optima AUC absorbance /interference centrifgue
- Max. RPM:50000
- No. of tubes accepted:8