ContactUsLinkComponent
GF Signet Installation Fitting, Weldolet Tee, Carbon Steel, 4" Pipe ID
  MFLX32511-80
undefined
GF Signet Installation Fitting, Weldolet Tee, Carbon Steel, 4" Pipe ID
  MFLX32511-80
  • Désignation:
    Installation Fitting, Weldolet Tee, Carbon Steel, 4" Pipe ID
  • Garantie:
    2 years
  • Cdt:
    1
  • Fitting material:
    Carbon Steel
  • Pipe size:
    4"
  • Process connection:
    Weldolet
Build a complete paddle-wheel flow sensor system customizable to your application.
  • Modular system for a customizable approach to flow measurement
  • Ideal for easy installation
The GF Signet paddle-wheel flow system is comprised of three components. Select the flow sensor, installation fitting, and flow meter that suit your application needs for a customizable approach to flow measurement.
Step One: Choose a Flow Sensor
Step Two: Choose an Installation Fitting
Step Three: Choose a Flow Meter
Flow sensor, installation fitting, and flow meter sold separately.
  • Désignation:
    Installation Fitting, Weldolet Tee, Carbon Steel, 4" Pipe ID
  • Environmentally Preferable:
  • Garantie:
    2 years
  • Cdt:
    1
  • Fitting material:
    Carbon Steel
  • Pipe size:
    4"
  • Process connection:
    Weldolet