Masterflex® I/P® Replacement Tubing Retainer Kit, PPS/PPS
Catalog # MFLX77602-06
Supplier: Avantor Fluid Handling
Specifications
- Pump series:I/P
- Désignation:I/P® Replacement Tubing Retainer Kit, PPS/PPS
- Garantie:3 Months
- Cdt:1
Specifications
About this item
Optimize performance of your I/P® Easy-Load® pump head.
- Constructed of polyphenylene sulfide (PPS)
Kit includes two polyphenylene sulfide (PPS) retainer assemblies, back plates, and assembly screws.
- Pump series:I/P
- Désignation:I/P® Replacement Tubing Retainer Kit, PPS/PPS
- Environmentally Preferable:
- Garantie:3 Months
- Cdt:1