Masterflex® Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 10/PK
Référence MFLX30600-23
Fournisseur: Avantor Fluid Handling
Spécifications
- Flow pattern:Three-Way
- Désignation:Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 10/PK
- Wetted materials:Polycarbonate
- Garantie:Out of box
- Cdt:10
- Inlet port:Female Luer
- Outlet port:Male Luer Lock
Spécifications
À propos de cet article
Enhance fluid flow with a larger bore than standard luer stopcocks.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Three-way flow pattern
Large-bore adapters are perfect for handling higher flow volumes in process while using connections similar to luer systems.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
- Flow pattern:Three-Way
- Désignation:Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 10/PK
- Environmentally Preferable:
- Wetted materials:Polycarbonate
- Garantie:Out of box
- Cdt:10
- Inlet port:Female Luer
- Outlet port:Male Luer Lock