Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pinces
Dumont #3c forceps
Dumont #3c forceps
Dumont #3c forceps
Catalog # FSCI11231-20
Supplier:
FINE SCIENCE TOOLS
Expand image
Dumont #3c forceps
Catalog # FSCI11231-20
Supplier:
FINE SCIENCE TOOLS
Specifications
Type:
#3c
Longueur:
110 mm
Matière:
Inox
Style de pointe:
Standard
Cdt:
1
Tip size:
0,17×0,1 mm
Tip shape:
Straight
Specifications
About this item
Product Details
Product Details