Dumont #6 forceps
#6, Inox, 115 mm, Standard, Angled, 0,17×0,1 mm
Catalog # FSCI11260-20
Marketsource
Supplier: FINE SCIENCE TOOLS
Jump to:
Product Details & Documents
Specifications
- Type:#6
- Longueur:115 mm
- Matière:Inox
- Style de pointe:Standard
- Tip size:0,17×0,1 mm
- Tip shape:Angled