0,00 SFr.
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Pinces
Dumont AA, polished inox forceps
Dumont AA, polished inox forceps
FSCI11210-20
:
FINE SCIENCE TOOLS
Dumont AA, polished inox forceps
FSCI11210-20
:
FINE SCIENCE TOOLS
Type:
AA
Longueur:
125 mm
Matière:
Inox
Style de pointe:
Standard
Cdt:
1
Tip size:
0,4×0,2 mm
Tip shape:
Straight