Vial forceps
Vial forceps
Acier inoxydable, 150 mm, Straight, 0,5 mm
Catalog # FSCI11006-15
Marketsource
Supplier:   FINE SCIENCE TOOLS
Vial forceps
Catalog # FSCI11006-15
Supplier:  FINE SCIENCE TOOLS
Marketsource
Vial forceps
Vial forceps
Acier inoxydable, 150 mm, Straight, 0,5 mm
Price
SFr. 128.71
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Longueur:
    150 mm
  • Matière:
    Acier inoxydable
  • Tip width:
    0,5 mm
  • Tip shape:
    Straight