ContactUsLinkComponent
Titanium bulldog spring clamp
Catalog # FSCI18144-30
Supplier:  FINE SCIENCE TOOLS
undefined
Titanium bulldog spring clamp
Catalog # FSCI18144-30
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Désignation:
    Light clamp press
  • Longueur:
    30 mm
  • Matière:
    Titane
  • Style de pointe:
    Serrated
  • Cdt:
    1
  • Tip size:
    9,5×1 mm
  • Tip shape:
    Straight