ContactUsLinkComponent
Dissector scissors, heavy blades
Dissector scissors, heavy blades
Catalog # FSCI14082-09
Supplier:  FINE SCIENCE TOOLS
undefined
Dissector scissors, heavy blades
Catalog # FSCI14082-09
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Type:
    Sharp/sharp, angled to side tips
  • Longueur:
    105 mm
  • Cdt:
    1
  • Longueur de lame:
    12 mm
Specifications
About this item
These scissors are made of stainless steel.