Dissector scissors, heavy blades
Catalog # FSCI14082-09
Supplier: FINE SCIENCE TOOLS
Specifications
- Type:Sharp/sharp, angled to side tips
- Longueur:105 mm
- Cdt:1
- Longueur de lame:12 mm
Specifications
About this item
These scissors are made of stainless steel.