Dust industrial PAPR kit, Jetstream®
Catalog # JSPACBP030-001-300
Supplier: JSP
Specifications
- Désignation:Jetstream® Dust industrial PAPR kit
- Cdt:1
Specifications
About this item
The Jetstream® Dust Industrial Kit consists of a belt-mounted rechargeable respirator with a large capacity disposable filter supplying clean air through a highly flexible reinforced hose to a grinding visor headtop.
- Grinding visor headtop
- No face fit required
- Flexible hose
- PSL dust filters
- Polycarbonate visor
- Multi-region plug adaptor
The Jetstream® Industrial kit contains: grinding visor headtop, flexible hose, PSL filters, flow meter, rechargeable battery, and multi-region plug adaptor.
Certifications: EN 12941; EN166.