Trace elements B, Corning®
N° de catalogue MDTC25-022-CI
Fournisseur: Corning
Numéro CAS:
Spécifications
- Cdt:100 mL
- Désignation:Trace elements B
Spécifications
À propos de cet article
Trace Elements B Solution has the same composition as Cleveland's Trace Element I.
The combination of Trace Elements B contains the trace elements of MCDB 104 and of MCDB 301.
This product is proprietary and not available from other major suppliers.