Anti-STAT5 (phospho-Tyr694) Mouse Recombinant Antibody [clone: A17016B.Rec] (PE (Phycoerythrin))
Fournisseur: Biolegend
Certificats
À propos de cet article
Spécifications
- Catalog No:
- BLEG936903
- BLEG936904
- Symbole de l'antigène:
- STAT5
- STAT5
- Synonymes antigène:
- Signal transducer and activator of transcription 5A
- Signal transducer and activator of transcription 5A
- Clonalité:
- Recombinant
- Recombinant
- Conjugaison:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- A17016B.Rec
- A17016B.Rec
- Réactivité:
- Human
- Human
- Hôte:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Cytométrie en flux:
- Yes
- Yes
- Modification:
- phospho-Tyr694
- phospho-Tyr694
- Immunogène:
- Peptide from Human Stat5A phosphorylated at Tyr694
- Peptide from Human Stat5A phosphorylated at Tyr694
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA)
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA)
- Température de stockage:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC
- ICFC
Spécifications
Frequently Bought Together