Anti-IFN-gamma Mouse Monoclonal Antibody [clone: B27] (Ultra-LEAF™)
Fournisseur: Biolegend
Certificats
À propos de cet article
Spécifications
- Catalog No:
- BLEG506532
- BLEG506533
- Symbole de l'antigène:
- IFN-gamma
- IFN-gamma
- Synonymes antigène:
- Type II interferon|T cell interferon|Macrophage-activating factor (MAF)|IFN-gamma|IFN-g|Interferon-γ|Immune interferon
- Type II interferon|T cell interferon|Macrophage-activating factor (MAF)|IFN-gamma|IFN-g|Interferon-γ|Immune interferon
- Clonalité:
- Monoclonal
- Monoclonal
- Conjugaison:
- Ultra-LEAF™
- Ultra-LEAF™
- Clone:
- B27
- B27
- Réactivité:
- Baboon, African green monkey, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human
- Baboon, African green monkey, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Pigtailed macaque, Chimpanzee, Human
- Hôte:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Cytométrie en flux:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Immunogène:
- E. coli expressed recombinant human IFN-γ
- E. coli expressed recombinant human IFN-γ
- Purification:
- The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- The Ultra-LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
- Storage buffer:
- 0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- 0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.01 EU/µg of the protein (<0.001 ng/µg of the protein) as determined by the LAL test.
- Température de stockage:
- 2…8 °C
- 2…8 °C
- Tested applications:
- ICFC, Neut, FC
- ICFC, Neut, FC
Spécifications
Frequently Bought Together