Anti-CD45RA Mouse Monoclonal Antibody [clone: HI100] (TotalSeq™-B0063)
Référence BLEG304161
Fournisseur: Biolegend
Spécifications
- Symbole de l'antigène:CD45RA
- Clonalité:Monoclonal
- Clone:HI100
- Conjugaison:TotalSeq™-B0063
- Hôte:Mouse
- Isotype:IgG2b kappa
- Réactivité:Chimpanzee,Human
- Synonymes antigène:PTPRC|GP180|T200|L-CA|LY5|LCA
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA.
- Température de stockage:2…8 °C
- Concentration:0.5 mg/ml
- Tested applications:PG
- Purification:The antibody was purified by chromatography and conjugated with TotalSeq™-B oligomer under optimal conditions.
- Taille:10 µg
- Cdt:10 µG