Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD95 (Fas) Mouse Monoclonal Antibody [clone: SA367H8] (PE (Phycoerythrin))
Anti-CD95 (Fas) Mouse Monoclonal Antibody [clone: SA367H8] (PE (Phycoerythrin))
Fournisseur:  Biolegend
Certificats
Anti-CD95 (Fas) Mouse Monoclonal Antibody [clone: SA367H8] (PE (Phycoerythrin))
Fournisseur:  Biolegend

Spécifications

  • Catalog No:
  • BLEG152608
  • BLEG152607
  • Symbole de l'antigène:
  • CD95 (Fas)
  • CD95 (Fas)
  • Synonymes antigène:
  • APO-1|ALPS1A|APT1|Fas
  • APO-1|ALPS1A|APT1|Fas
  • Clonalité:
  • Monoclonal
  • Monoclonal
  • Conjugaison:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • SA367H8
  • SA367H8
  • Réactivité:
  • Mouse
  • Mouse
  • Hôte:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Cytométrie en flux:
  • Yes
  • Yes
  • Immunogène:
  • Mouse Fas Transfectants
  • Mouse Fas Transfectants
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
  • Concentration:
  • 0.2 mg/ml
  • 0.2 mg/ml
  • Storage buffer:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Température de stockage:
  • 2…8 °C
  • 2…8 °C
  • Tested applications:
  • FC
  • FC

Spécifications

Frequently Bought Together