Anti-CD45 Mouse Monoclonal Antibody [clone: HI30] (APC (Allophycocyanin)/FIRE™750)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG304061
- BLEG304062
- Symbole de l'antigène:
- CD45
- CD45
- Synonymes antigène:
- T200|LCA
- T200|LCA
- Clonalité:
- Monoclonal
- Monoclonal
- Conjugaison:
- APC (Allophycocyanin)/FIRE™750
- APC (Allophycocyanin)/FIRE™750
- Clone:
- HI30
- HI30
- Réactivité:
- Chimpanzee, Human
- Chimpanzee, Human
- Hôte:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Cytométrie en flux:
- Yes
- Yes
- Purification:
- The antibody was purified by affinity chromatography and conjugated with APC/Fire™ 750 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with APC/Fire™ 750 under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Température de stockage:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together