Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG368504
- BLEG368503
- Symbole de l'antigène:
- CD45
- CD45
- Synonymes antigène:
- T200|Leukocyte Common Antigen (LCA)
- T200|Leukocyte Common Antigen (LCA)
- Clonalité:
- Monoclonal
- Monoclonal
- Conjugaison:
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Clone:
- 2D1
- 2D1
- Réactivité:
- Human
- Human
- Hôte:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Cytométrie en flux:
- Yes
- Yes
- Immunogène:
- Human PBMC
- Human PBMC
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PerCP/Cyanine5.5 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PerCP/Cyanine5.5 under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Température de stockage:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together