QUANTOFIX Iron Sensitive 1
N° de catalogue MANA91359
Fournisseur: MACHEREY-NAGEL
Numéro CAS:
Spécifications
- Cdt:25
- Désignation:QUANTOFIX Iron Sensitive 1, 25 tests
Spécifications
À propos de cet article
Semi-quantitative test strips for reliable determination of iron concentrations as low as 0.05 mg/L (ppm). A hole in the plastic carrier allows the water to migrate through the test pad increasing the sensitivity by a factor of nearly 10. Ideal for rapid and easy analysis of iron directly on-site. No maintenance or calibration is required.
- Sensitive rapid test