Bottles, amber, glass, with thread, Assistent®
N° de catalogue HECH40190030
Fournisseur: GLASWARENFABRIK KARL HECHT
Spécifications
- Capacité:1000 ml
- Couleur:ambré
- Désignation:Glass bottle with thread
- Seq:02
- Environmentally Preferable:
- Cdt:1
Spécifications
À propos de cet article
Amber glass finish is designed to protect light-sensitive samples in transport and storage.
- Space saving square design
- With screw cap
Bottles are supplied with a screw cap made of polypropylene, which is resistant to steam sterilsation up to 121 °C (249,8 °F).