StrataPrep Plasmid Purification kits
StrataPrep Plasmid Purification kits
StrataPrep plasmid miniprep kit, 1
N° de catalogue AGLS400763
Fournisseur:   AGILENT
StrataPrep Plasmid Purification kits
N° de catalogue AGLS400763
Fournisseur:  AGILENT
Numéro CAS:  
StrataPrep Plasmid Purification kits
StrataPrep plasmid miniprep kit, 1
Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
The StrataPrep plasmid preparation kits provide a rapid method for purifying plasmid DNA from bacterial cultures.
  • High yields of plasmid DNA: up to 20 µg from 1,5 to 3,0 ml of culture
  • Accommodates all sizes of DNA from standard 2,9 kb plasmids to 45 kb cosmids
  • Transforms as efficiently as cesium chloride-purified DNA
The kit utilises a special silica-based fibre matrix technology in a microspin-column format to capture DNA in the presence of a chaotropic salt, allowing the recovery of exceptionally clean DNA.
Contains: Solution 1, 2, 3, Wash buffer, Nuclease-removal buffer, uspin cups and receptacle tubes.
Attention: For research use only. Not for use in diagnostic procedures.
Spécifications
Spécifications
  • Cdt:
    1
  • Désignation:
    StrataPrep plasmid miniprep kit
  • Cat. No.:
    AGLS400763
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Target molecule:
    Plasmid
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Bacteria