Order Entry
Switzerland
ContactUsLinkComponent
N-Boc-Glycylglycylglycine
N-Boc-Glycylglycylglycine
Catalog # APOSOR471623-25G
Supplier:  Apollo Scientific
CAS Number:  
N-Boc-Glycylglycylglycine
Catalog # APOSOR471623-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Cdt:
    25 g
  • Nom de la protéine/du peptide:
    N-Boc-Glycylglycylglycine
  • Purity:
    0.95

Specifications

About this item