Rat EGF
- Catalog No:
- APOSBITP2077-2UG
- APOSBITP2077-5UG
- APOSBITP2077-10UG
- Nom de la protéine/du peptide:
- EGF
- EGF
- EGF
- Synonyme de protéine:
- Epidermal Growth Factor
- Epidermal Growth Factor
- Epidermal Growth Factor
- Species:
- Rat
- Rat
- Rat
Product Family Options
- Cdt
Specifications
Cat. No.APOSBITP2077-2UGNom de la protéine/du peptideEGFSynonyme de protéineEpidermal Growth FactorSpeciesRatSpecifications
Cat. No.APOSBITP2077-5UGNom de la protéine/du peptideEGFSynonyme de protéineEpidermal Growth FactorSpeciesRatSpecifications
Cat. No.APOSBITP2077-10UGNom de la protéine/du peptideEGFSynonyme de protéineEpidermal Growth FactorSpeciesRat