Human Recombinant PDGF-BB
- Catalog No:
- APOSBITP1235-2UG
- APOSBITP1235-10UG
- Nom de la protéine/du peptide:
- PDGF-BB
- PDGF-BB
- Synonyme de protéine:
- Platelet Derived Growth Factor-BB
- Platelet Derived Growth Factor-BB
- Type de protéine/peptide:
- Recombinant
- Recombinant
- Species:
- Human
- Human
Product Family Options
- Cdt
Specifications
Cat. No.APOSBITP1235-2UGNom de la protéine/du peptidePDGF-BBSynonyme de protéinePlatelet Derived Growth Factor-BBType de protéine/peptideRecombinantSpeciesHumanSpecifications
Cat. No.APOSBITP1235-10UGNom de la protéine/du peptidePDGF-BBSynonyme de protéinePlatelet Derived Growth Factor-BBType de protéine/peptideRecombinantSpeciesHuman