SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Protéines et Peptides
Mouse Recombinant IGF-I
Mouse Recombinant IGF-I
APOSBITP1107-1MG
:
Apollo Scientific
:
Mouse Recombinant IGF-I
APOSBITP1107-1MG
:
Apollo Scientific
:
Cdt:
1 mg
Type de protéine/peptide:
Recombinant
Species:
Mouse
Synonyme de protéine:
Insulin Like Growth Factor-I
Nom de la protéine/du peptide:
IGF-I