NANOCOLOR® Kit de test en tube photométrique pour hydrocarbures
A pre-dosed NANOCOLOR® tube test for photometric determination of hydrocarbons in water.
Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
: Rugged box with 20 test tubes.
- Modèle:MANA985057
- Plateforme:NANOCOLOR® tube tests
- Température de stockage:15…25 °C
- Durée de conservation (mois):12
- Poids:460 g (including packaging)
Frequently Bought Together