PIG® eco-friendly compact spill kit
Catalog # 121-0191
Anbieter: NEW PIG
Technische Daten
- Désignation:Spill kit, incl 4 mat pads, 1 pillow and 2 disposal bags
- l×p×h:100×380×460 mm
- Environmentally Preferable:Sustainable Materials
- Capacité d'absorption:7 L
- Cdt:1 KIT
Technische Daten
Über diesen Artikel
This compact eco-friendly kit makes transport simple, and its recycled content pads and pillow soak up more liquid with less environmental impact.
- Environmentally friendly absorbents feature high recycled content; reuses material that would normally enter a waste stream
- Reusable wallet is constructed out of jute plant bark; naturally strong and durable with waterproof inner coating to keep the kit clean, dry and ready to use
- High visibility colour helps identify spill kit location; saves valuable time during response
- ‘Hook and loop’ closure is easy to open for fast access to absorbents
- Carrying handle affords convenient transport; makes it ideal for hanging on a wall, storing in a cabinet or being carried in a vehicle cab
- Compact design takes up little storage room and effectively deals with minor spillages
Super absorbent PIG BLUE® mat pads and PIG® pillow have maximum capacity for soaking up spills of oils, coolants, solvents or water.