PIG® Bio-fluids solidifying absorbent powder
N° de catalogue NEWPPLP503
Fournisseur: NEW PIG
Spécifications
- Désignation:Bio-fluids solidifying absorbent powder
- Capacité d'absorption:30,3 L/container
- Emballage:4 containers per box
- Cdt:4 Bottle
Spécifications
À propos de cet article
Fast-acting absorbent captures biofluids like blood, urine and vomit and "locks" them inside for cleaner, safer disposal.
- Permanently solidifies to lock in absorbed liquids so they won't leak or liquefy when the absorbents are handled or placed under pressure
- Contains chlorine to kill pathogens and reduce odors
- Clean scent helps reduce offensive odors during and after cleanup
- Environmentally-friendly product absorbs biological fluids including blood, urine and vomit
- Forms a solid, non-expanding mass for easier, less-expensive recovery and cleanup
- Shaker bottle makes it easy to apply absorbent directly onto spill area
High Btu value produces less than 0.1% ash content, ideal for fuels blending and incineration.