AccuScript high-fidelity reverse transcriptase
AccuScript high-fidelity reverse transcriptase
Fournisseur:   AGILENT
AccuScript high-fidelity reverse transcriptase
Fournisseur:  AGILENT
Certificats
AccuScript high-fidelity reverse transcriptase
2 variantes
Product Details & Documents
AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.
  • MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
  • High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0,1 to 10 ng of poly(A)+ RNA
  • Stringent quality control procedures ensure high yields of premium cDNA
AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3,7 times more accuracy than other commercially available RTs and 6,6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Details
    No. of reactions
    Cdt
    Disponibilité
    Prix
  • AccuScript reverse transcriptase
    Engineered to deliver the highest transcription accuracy while promoting full-length cDNA synthesis, yield, and RT-PCR performance
    50 reactions
    1
    Connexion Pour la disponibilité
    Connexion Pour la disponibilité
    1 * 1 ST
    1 * 1 ST
  • High fidelity reverse transcriptase for long target lengths. Ideal for low RNA input and high specificity. Shorter PCR extension times with PfuUltra II with high accuracy. Kit includes dNTP, dTT, oligodT and random primers. Capable of amplifying cDNA of 0,1 - 9,6 kb in length ;10 - 1000 ng of total RNA or 0,1 - 10 ng of poly(A)+ RNA input
    50 reactions