Molecular detection matrix control
Contrôle de la matrice de détection moléculaire, 96
N° de catalogue MMMPMDMC96NA
Fournisseur: 3M Food Safety
Molecular detection matrix control
N° de catalogue MMMPMDMC96NA
Fournisseur: 3M Food Safety
Numéro CAS:
Aller à
Product Details & Documents
Kit contains 96 tests and is used with the 3M™ Molecular Detection System to check for inhibition of the matrix sample during validation periods, when adopting the 3M™ method or testing new or unknown matrices.
- Verify that new or unknown matrices are not inhibitory to amplification
- Ready-to-use and pre-dispensed reagents
- Only two transfer steps after enrichment
- Closed-tube system reduces risk of amplicon contamination
- Designed to work with 3M™ proprietary enrichment broths to promote growth
Spécifications
Spécifications
- Cdt:96
- Désignation:Contrôle de la matrice de détection moléculaire