Molecular detection assay E. coli O157
Catalog # MMMPMDA2ECO96
Supplier: 3M Food Safety
Specifications
- Désignation:E.coli 96 tests/kit
- Cdt:96 Tests
Specifications
About this item
3M™ Molecular Detection Assay 2 - E. coli O157 (including H7) is used with the 3M™ Molecular Detection System for the rapid and specific pathogen detection of E. coli O157, including H7, in enriched food samples.
- One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees
- Ready-to-use and pre-dispensed reagents provide convenience and minimise the chance of human error
- Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification
- The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently
- Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster
As with all test methods, the source, formulation and quality of enrichment medium can influence the results. 3M has evaluated the 3M Molecular Detection Assay 2 E. coli O157 (including H7) with Buffered Peptone Water ISO.