ClonePix™ 2 Mammalian Colony Picker
Référence MLDVX4960/220
Fournisseur: Molecular Devices
Spécifications
- Désignation:ClonePix 2 system configures for 220V. Includes self contained Class 100 environmental unit, imaging station, picking head, dedicated PC with software pre-loaded. 1 year warranty covering parts and labor Compressor not included (Compatible with optional standalone compressor)
- Cdt:1
Spécifications
À propos de cet article
Redefine clone screening and selection: transform your cell line development workflow.
- Screen more clones in less time
- Accurate, automatic colony picking avoids errors associated with limiting dilution
- Excellent image quality allows for screening of stable, high-producing clones
- Increased productivity of a cell line development workflow
- Add FL and white light imaging - pull bullets from key specs
The ClonePix™ 2 Mammalian Colony Picker is a fully automated system for the selection of high-value clones used in antibody discovery and cell line development. Hybridoma, CHO cells, and other cell types are imaged and selected based on user-defined parameters. Plate handling, barcode reading, and picking are all fully integrated. All data, including images, are saved for downstream analysis. The picker increases the probability of finding optimally produced cell lines and significantly reduces time and labor.