Hand towels, folded, HOSTESS*
Catalog # KIMB6876
Supplier: KIMBERLY CLARK
Specifications
- Emballage:12 sleeves of 180 towels
- Couleur:bleu
- Désignation:Paper towels, 1 ply
- L×l:240×230 mm
- Seq:03
- Cdt:2.688
Specifications
About this item
Made of 100% recycled fibre, these paper towels open more easily and are dispensed one at a time so they are touched only by the user.
- Helps to dry hands hygienically and effectively
- Help reduce costs and waste
Ideal for busy washroom environments including schools and other public facilities where economy is important.