Cleanroom wipes, pre-saturated, alcohol, Kimtech™ W4
Catalog # KIMB06070
Supplier: KIMBERLY CLARK
Specifications
- Emballage:400 lingettes/boîte
- L×l:228,6×279,4 mm
- Cdt:400
Specifications
About this item
The 100% polypropylene wipers come pre-saturated with 70% isopropyl alcohol (semiconductor grade) and 30% deionized water, in a resealable pouch.
- Ideal one-step application of alcohol to critical surfaces
- Low lint for environments where 'lint free' standards are important
- These disposable wipers are recommended for ISO Class 4 and higher clean rooms
These disposable wipers are ideal for use in pharmaceutical, high-tech, biotechnology and medical-device industries that require alcohol wiping for critical tasks.
Certifications: Meets EPA guidelines for minimum post consumer waste content.
Packaging: Package containe 40 wipes/pack, 10 packs/case, 400 wipes/case.
Caution: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904