Chemical resistant gloves, G80, KleenGuard™
Fournisseur: KIMBERLY CLARK
Certificats
À propos de cet article
KleenGuard™ G80 Neoprene/latex chemical resistant gloves used in chemical protection, comfort and durability.
- Removing paint, sealants or residues from a surface or degreasing machinery with a strong solvent
- Cotton flock linking, which adds comfort and absorbs perspirations for easy donning and removal
They have a natural rubber latex construction, covered in neoprene, which gives them high resistance to acids, alkalis, oils and solvents. They are a smart choice for cleaning and degreasing, solvent wiping for parts and machinery, handling acids, bases, alcohols, alkalis, phenols and salts and general chemical processing.
Certifications: EN 374-AKL
Frequently Bought Together