Sand baths, SB400E
Bain de sable, SB400E, 320×320 mm
N° de catalogue 453-0083
Fournisseur: SUMINISTROS GRUPO ESPER S L
Aller à
Product Details & Documents
Digital stainless steel sand bath, controlled by microprocessor. Homogenic distribution of heath for uniform heating of the complete contact surface.
- Temperature controlled by microprocessor, with internal PT100 sensor
- 'ARV V2' fuzzy logic algorithm for high precision temperature control, to be used with external sensor, without surpassing reference temperature (even at 30 °C)
- Digital display LCD backlighted showing the selected and actual temperature, and timer (0 to 120 min) with audible alarm
- Indication on the display of high temperature on the surface, that protects against burn
- Automatic restart after unexpected shut down keeping the same parameters as before the incident. An incident sigh appears on the display
- Membrane push-button control
Certifications: IP Protection class: IP 52
Spécifications
- Désignation:Bain de sable, SB400E
- Dimensions de la plaque:320×320 mm
- Cat. No.:453-0083
- Poids:7,5 kg
- Gamme de température:Température ambiante +5…400 °C
- L×P×H:360×460×180
- Puissance:1500 W
- Modèle:SB400E