Chemical resistant gloves, Nitrosafe Plus, A812
: Portwest
This nitrile gauntlet is tested and certified to offer protection against many common chemicals found in industry today.
- Anti-slip pattern offers high dexterity in wet and dry conditions
- Ultra-sensitive and free of soluble proteins help minimise the risk of allergic reaction
- Textured pattern for enhanced grip
- Flock lined for added comfort
Ideal for manufacturing, paint applications, electronics and glass handling where silicone is problematic. Suitable for a broad range of industries, including industrial cleaning, laboratory work, light assembly, aerospace and the chemical industry.
: EN ISO 374-5 micro organisms; EN 388:2016 (3101X); EN ISO 374-1:2016 Type A (AJKLMNOPST); Cat. III
Food contact approved
Food contact approved
- Catalog No:
- PRTWA812GNRXS
- PRTWA812GNRXXL
- PRTWA812GNRS
- PRTWA812GNRXL
- PRTWA812GNRL
- PRTWA812GNRM
- Ambidextre/anatomique:
- Anatomiques
- Anatomiques
- Anatomiques
- Anatomiques
- Anatomiques
- Anatomiques
- Usage unique/Réutilisable:
- réutilisable
- réutilisable
- réutilisable
- réutilisable
- réutilisable
- réutilisable
- Matériau du support:
- Coton
- Coton
- Coton
- Coton
- Coton
- Coton
- Finition:
- Textured
- Textured
- Textured
- Textured
- Textured
- Textured
- Style de manchette:
- À crispin
- À crispin
- À crispin
- À crispin
- À crispin
- À crispin
- Norme EN:
- EN ISO 374-5; EN 388:2016 (3101X); EN ISO 374-1:2016 Type A (AJKLMNOPST)
- EN ISO 374-5; EN 388:2016 (3101X); EN ISO 374-1:2016 Type A (AJKLMNOPST)
- EN ISO 374-5; EN 388:2016 (3101X); EN ISO 374-1:2016 Type A (AJKLMNOPST)
- EN ISO 374-5; EN 388:2016 (3101X); EN ISO 374-1:2016 Type A (AJKLMNOPST)
- EN ISO 374-5; EN 388:2016 (3101X); EN ISO 374-1:2016 Type A (AJKLMNOPST)
- EN ISO 374-5; EN 388:2016 (3101X); EN ISO 374-1:2016 Type A (AJKLMNOPST)
- Convient au contact alimentaire:
- +
- +
- +
- +
- +
- +
Frequently Bought Together