Handheld refractometer for ethyl alcohol, PAL-Covid-19
Catalog # ATAG4553
Supplier: Atago
Specifications
- Désignation:Ethyl alcohol refractometer, PAL-Covid-19
- Cdt:1
Specifications
About this item
PAL-Covid19 will measure the concentration of alcohol percentage for hand made sanitizer.
- Measure ethanol concentration easily
You can easily measure the alcohol concentration with PAL-Covid19 in 4 steps. Low alcohol concentration is ineffective for sterlization, but high alcohol conentration may cause rough skin.
Ordering information: Requires 2 AAA batteries.