Cable ties, stainless steel
Katalognr. BRDY134057
Supplier: Brady
Specifications
- Couleur:argenté
- Désignation:Cable ties
- Dimensions:4,5×127 mm
- Cdt:100
Specifications
About this item
Brady B-7316 stainless steel cable ties are made from 316 grade stainless steel and are designed to withstand harsh environments, and offer excellent outdoor durability.
- Thickness of 0,5 mm