Labels for BBP33 printers, type B-472
À propos de cet article
Flame retardant wire wraps (B-472) are ideal for cable and wire bundle applications and label applications where self-extinguishing properties are required.
- Polyimide material specifically designed to self-extinguish and prevent flames from propagating
- Service temperature of –40 to 100 °C
Matte finish material with permanent acrylic adhesive can be thermal transfer printed or dot matrix printed.
Certifications: RoHS approved.
11 variantes
Product Details & Documents
Flame retardant wire wraps (B-472) are ideal for cable and wire bundle applications and label applications where self-extinguishing properties are required.
- Polyimide material specifically designed to self-extinguish and prevent flames from propagating
- Service temperature of –40 to 100 °C
Matte finish material with permanent acrylic adhesive can be thermal transfer printed or dot matrix printed.
Certifications: RoHS approved.
Product Family Options
Labels
Informations sur le produit
- DésignationTypeModèleMatièreAdhésifCouleurPrinting methodImprimeurDimensionsEmballageDisponibilitéPrix
- Labels, topcoated polyimideB-472B33-11-472PolyimidePermanent, acryliqueblancBBP336,4×6,4 mm12 labels/rowConnexion Pour la disponibilité1 * 15.000 STSFr. 181.00
- Labels, topcoated polyimideB-472B33-1-472PolyimidePermanent, acryliqueblancBBP3319,1×6,4 mm4 labels/rowConnexion Pour la disponibilité1 * 5.000 STSFr. 181.00
- Labels, topcoated polyimideB-472B33-2-472PolyimidePermanent, acryliqueblancBBP3322,9×6,4 mm3 labels/rowConnexion Pour la disponibilité1 * 5.000 STSFr. 217.00
- Labels, topcoated polyimideB-472B33-5-472PolyimidePermanent, acryliqueblancBBP3325,4×12,7 mm3 labels/rowConnexion Pour la disponibilité1 * 5.000 STSFr. 481.00
- Labels, topcoated polyimideB-472B33-5-472YLPolyimidePermanent, acryliquejauneBBP3325,4×12,7 mm3 labels/rowConnexion Pour la disponibilité1 * 5.000 STSFr. 481.00
- Labels, topcoated polyimideB-472B33-29-472PolyimidePermanent, acryliqueblancBBP3331,8×9,5 mm2 labels/rowConnexion Pour la disponibilité1 * 5.000 STSFr. 481.00
Labelling tapes
Informations sur le produit
- DésignationTypeModèleMatièreAdhésifCouleurPrinting methodImprimeurDimensionsEmballageDisponibilitéPrix
- Topcoated label printer tapeB-472B33C-1000-472PolyimidePermanent, acryliqueblancThermal transferBBP3325,4 mm×25,9 m1 roll/boxConnexion Pour la disponibilité1 * 1 STSFr. 204.00
- Topcoated label printer tapeB-472B33C-1000-472YLPolyimidePermanent, acryliquejauneThermal transferBBP3325,4 mm×25,9 m1 roll/boxConnexion Pour la disponibilité1 * 1 STSFr. 204.00
- Topcoated label printer tapeB-472B33C-2000-472PolyimidePermanent, acryliqueblancThermal transferBBP3350,8 mm×25,9 m1 roll/boxConnexion Pour la disponibilité1 * 1 STSFr. 321.00
- Topcoated label printer tapeB-472B33C-2000-472YLPolyimidePermanent, acryliquejauneThermal transferBBP3350,8 mm×25,9 m1 roll/boxConnexion Pour la disponibilité1 * 1 STSFr. 321.00
- Topcoated label printer tapeB-472B33C-3000-472YLPolyimidePermanent, acryliquejauneThermal transferBBP3376,2 mm×25,9 m1 roll/boxConnexion Pour la disponibilité1 * 1 STSFr. 439.00