Sign, message kit, A4
Message sign kit, A4, Plastique, 210×297 mm
Catalog # BRDY292250
Supplier: Brady
Jump to:
Product Details & Documents
Create an even safer work environment by using these universal signs.
- The plastic frame simply clips to the posts
Delivery information: Comes as a complete kit: frame with two attachment clips and one chain (200 mm).
Specifications
- Désignation:Message sign kit, A4
- Dimensions:210×297 mm
- Matière:Plastique