2-Methylnaphtho[2,3-d][1,3]thiazol ≥98.0% (par GC)
TCIAM1287-5G
: TCI
:
- Cdt:5 g
- Seq:3173
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:94.0 to 97.0 deg-C
- Solubility in Acetonitrile:almost transparency
:
:168°C
:199.27 g/mol
:220-240-1
:2682-45-3
:C₁₂H₉NS
:MFCD00004951