ContactUsLinkComponent
4-(Hydroxymethyl)phenoxymethyl polystyrene resin 0,5 - 1,0 mmol/g, "cross-linked" 1% DVB (200-400 mesh)
  TCIAH0986-5G
 :  TCI
 :  
undefined
4-(Hydroxymethyl)phenoxymethyl polystyrene resin 0,5 - 1,0 mmol/g, "cross-linked" 1% DVB (200-400 mesh)
  TCIAH0986-5G
 :  TCI
 :  
  • Cdt:
    5 g
  • Seq:
    3173
  • Loading:
    0.5 to 1.0 mmol/g
 :
 :MFCD00131778