USB camera, Moticam® S1
Référence 630-3099
Fournisseur: MOTIC
Spécifications
- Modèle:Moticam S1
- Résolution:1,2 MP
- Type de sonde:sCMOS
- Format de sonde:1/3"
- Fréquence de trame:1280×960 @ 120 fps; 640×480 @ 240 fps, max.
- Systèmes d'exploitation:Windows 7/8/10, MAC OSX 10.9, Linux or higher
- Montage de lentille:C-mount
- Support de compatibilité:TWAIN, SDK and DirectShow Driver
- Interfaces:USB 3.1
- Sonde:sCMOS
- Logiciel:Motic Images Plus 3.0 for Windows, OSX and Linux
- Cdt:1
Spécifications
À propos de cet article
High Grade sCMOS USB camera Moticam® S1 for microscopy for an ultimate imaging result.
- Fast frame rates
- sCMOS sensors with back-illuminated sensor technology
- USB 3.1 interface
- Motic Images Plus 3.0 software
Moticam S1 is an the entry model for back-illuminated sensor technology and speedy data transfer (with frame rates up to 120 fps*), with easy image setting and capturing. Create your perfect images for a professional database.
Informations de commande: To be purchased separately – C-mount adapter not included, this needs to the same brand as your microscope, compatible with your microscope model and fitting the size of your Moticam chip.
Livraison: Includes CS ring adapter, USB 3.1 cable, Motic 4-dot calibration slide, Motic Images Plus 3.0 for Windows/OSX/Linux.
Attention: Minimum computer requirements – 2 GHz dual-core, RAM memory 2 GB, video memory min. 512 MB.