Reverse osmosis water purification system, Millipore®
Module d'osmose inverse
172-5087
: Merck
This Reverse Osmosis membrane removes 95 to 99% of contaminants including ions, particulates, bacteria, and organic molecules (MW >200kDa)
Height: 41 cm
- Désignation:Module d'osmose inverse
- Cat. No.:172-5087
- Débit:5 L/h