Avantor® Hichrom, UHPLC Column Connector
Catalog # HICHHI-903
Supplier: Avantor
Specifications
- Désignation:UHPLC connector for 1/16" O.Ø tubing
- Cdt:1
Specifications
About this item
This reusable UHPLC column connector have 10-32 threads. This fitting is reusable, as they are not permanently swaged onto the inlet tubing.
- Compatible with all UHPLC systems and column brands
- Avoids poor connections
- Pressure rated to >1700 bar (>25000 psi)
An internal ferrule design prevents tubing slippage, whilst the PEEK-polymer blend front ferrule eliminates the risk of damage to receiving ports.